CRITERION™ Thiogelatin Medium
Supplier: HARDY DIAGNOSTICS
C8240 C8241
89406-734EA 169.11 USD
89406-734 89406-736
CRITERION™ Thiogelatin Medium
Media for Microbiology Prepared Media for Microbiology
CRITERION™ Thiogelatin medium is intended to be used for the differentiation of microorganisms based on their ability to liquefy gelatin.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certificaciones: BSE Free.