CRITERION™ Peptone Water
Supplier: HARDY DIAGNOSTICS
C6570 C6571 C6572 C6573
89405-866EA 54 USD
89405-866 89405-868 89405-870 89405-872
CRITERION™ Peptone Water
Media for Microbiology Dehydrated Media for Microbiology
CRITERION™ peptone water is used for the cultivation of nonfastidious organisms, for performing the indole test, and for studying fermentation patterns.
  • Wide mouth opening - reduces dust and allows easy scooping
  • Convenient handgrip - 'no slip' finger holds
  • Stackable containers
  • High solubility - dissolves quickly
  • Custom and bulk products available - to your specifications
Dehydratedculture media with engineered container.
Certificaciones: BSE Free.