Oxford Listeria Agar Base, CRITERION™, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6520
C6521
C6522
C6523
89405-824EA
92.96
USD
89405-824
89405-826
89405-828
89405-830
Oxford Listeria Agar Base, CRITERION™, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CRITERION™ Oxford Listeria Agar Base is used for the selective isolation of Listeria moncytogenes from food
- Wide mouth opening - reduces dust and allows easy scooping
- Convenient handgrip - 'no slip' finger holds
- Stackable containers
- High solubility - dissolves quickly
- Custom and bulk products available - to your specifications
Listeria Selective Supplement, product #SR140E, is required for this medium. Dehydratedculture media with engineered container.
Certificaciones: BSE Free