LB Broth, Miller, Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C6010
C6011
C6012
C6013
C6019
89405-570EA
56.37
USD
89405-570
89405-572
89405-574
89405-576
89405-578
LB Broth, Miller, Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology
Dehydrated Media for Microbiology
CulGenex™ LB Broth, Miller, is used for the maintenance and propagation of Escherichia coli used in molecular biology procedures.