CulGenex™ LB Broth, Lennox, Dehydrated Culture Media, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
C7650 C7651 C7652 C7653
89406-468EA 55.34 USD
89406-468 89406-470 89406-472 89406-474
CulGenex™ LB Broth, Lennox, Dehydrated Culture Media, Hardy Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
CulGenex™ LB Broth, Lennox is recommended for maintaining and cultivating recombinant strains of Escherichia coli for molecular biology procedures
CulGenex™ Dehydrated Culture Media is formulated to meet or exceed the highest standards. Each lot of CulGenex™ media is meticulously tested for performance and reproducibility.
Información para pedidos: CulGenex™ LB Broth, Lennox is available in 2 L mylar puches, pre-measured for 2 liter patches; 500 mL mylar puches, pre-measured for 500 mL batches with 6 pouches per pack; 500 g bottles with wide mouth, easy grip and scoop included; 2 kg and 10 kg buckets with screw-on lids.