The Thermo Scientific Autostainer 360 Stainer and Autostainer 720 Stainers deliver flexibility and productivity for immunohistochemistry automation. Multiple sizes are available to accommodate the needs of nearly any size laboratory. For maximum efficiency and reproducibility with formalin fixed samples, using the Autostainer and PT Module together as a system is recommended.
The Autostainer and PT Module Series offer proven reliability along with the flexibility and value of a completely open system
The Thermo Scientific PT Module antigen retrieval device precisely duplicates antigen retrieval conditions across all slides. The gentle heat quickly produces outstanding results while maintaining the integrity of delicate tissue. In addition, most users can streamline testing practices by consolidating various antigen retrieval methods into only one or two PT Module antigen retrieval device procedures, which can be run simultaneously. Precise temperature control without the damaging effects of temperature variations, microwaves, or pressure delivers a level of consistency unmatched by any other instrument available.
System features and benefits include, full regulatory compliant reporting and data management, efficiency with pretreatment through counterstain automation, error reduction with dual bar coding (slides and reagents), and enhanced lean workflow with on-demand labeling directly to slides. The interface can be configured for diagnostic or research applications at no additional cost.
Información para pedidos: Autostainer 360 and Autostainer 720 systems include 36-slide Autostainer/84-slide Autostainer with 2D matrix bar code reader, computer, monitor, label printer(s), document printer, 3 metal reinforced ABS plastic slide racks, reagent rack, reagent vials and caps, autostainer probe cleaning reagent kit, buffer rinse bottle, wash buffer, DI water, waste containers, and universal power supply.