Z-Leu-Leu-Glu-Amc
Supplier: Enzo Life Sciences
ZW9345-0005 ZW9345-0050
89165-662EA 240.89 USD
89165-662 76045-560
Z-Leu-Leu-Glu-Amc
Proteins and Peptides
Fluorogenic substrate for the peptidylglutamyl-peptide hydrolysing (caspase-like) activity of the proteasome which may be stimulated in the presence of Mg2+ ions and rapidly inactivated by N-acetylimidazole.