VU0155069 ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: (S)-N-(1-(4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidin-1-yl)propan-2-yl)-2-naphthamide

BML-ST370-0005 BML-ST370-0025
89164-830EA 130.8 USD
89164-830 89164-832
VU0155069 ≥98% (by TLC)
VU0155069

PLD inhibitor


Potent and specific inhibitor of phospholipase D1 (PLD1) in cellular (PLD1 IC50 = 11nM Calu-1 cells; PLD2 IC50 = 1800nM HEK293-gfpPLD2 cells) and in vitro (hPLD1 IC50 = 46nM; hPLD2 IC50 = 933nM) assays. Significantly decreases invasive migration in breast cancer cell lines (MDA-231, 4T1 and PMT).

Formula: C₂₆H₂₇ClN₄O₂
MW: 462.970 g/mol
MDL Number: MFCD23160355
CAS Number: 1130067-06-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2afb982827a66b6096470732f092b6c3bad14547&vid=9afb32822a42afe34e785efdb07f7c7e9c80ec97&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR