Methyl arachidonyl fluorophosphonate ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-ST360-0005 BML-ST360-0025
89164-826EA 170.04 USD
89164-826 89164-828
Methyl arachidonyl fluorophosphonate ≥98% (by TLC)
Methyl arachidonyl fluorophosphonate

Phospholipase inhibitor


Selective, active-site directed, irreversible inhibitor of cPLA2 (IC50=5µM) and Ca2+-independent iPLA2 (IC50=0.5µM). Potent, irreversible inhibitor of fatty acid amide hydrolase (FAAH) (IC50=2.5nM), 800 -fold more potent than AACOCF3 (Prod. No. BML-ST335)). Irriversible CB1 receptor antagonist in rat brain membrane (IC50=20nM).

Formula: C₂₁H₃₆FO₂P
MW: 370.489 g/mol
MDL Number: MFCD03095722
CAS Number: 188404-10-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=17b831aee64a029b9c71fc2b091cdef2f4a03122&vid=168b5123916ddb14c8663a0d163ad63791174671&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR