AACOCF3 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-ST335-0010 BML-ST335-0050
89164-806EA 163.5 USD
89164-806 89164-808
AACOCF3 ≥98% (by TLC)
AACOCF3

AACOCF3 is a trifluoromethylketone derivative of arachidonic acid. It is a selective inhibitor of human cytosolic PLA2 and Ca2+-independent PLA2 (iPLA2, IC50=15 µM). It is active in intact platelets and U937 cells (IC50=8 µM). It also inhibits fatty acid amide hydrolase (100% at 7.5 µM).


  • Inhibitor of PLA2 and FAAH


Yellow oil.


Soluble in DMSO (>25mg/ml) or 100% ethanol (>25mg/ml).

Formula: C₂₁H₃₁F₃O
MW: 356.5 g/mol
Storage Temperature: Ultra-Low Temperature Freezer (–80°C)
CAS Number: 149301-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR