Ono-RS-082 ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-ST320-0020 BML-ST320-0100
89164-794EA 107.91 USD
89164-794 89164-796
Ono-RS-082 ≥98% (by TLC)
Ono-RS-082

Phospholipase A2 inhibitor (IC50=7 µM for purified guinea pig lung PLA2). Inhibits epinephrine-stimulated thromboxane production in human platelets (86% at 3.5µM). Also blocks insulin secretion in pancreatic islets in response to glucose. Disrupts membrane tubule-mediated reassembly and maintenance of the Golgi complex.


  • Phospholipase inhibitor


Off-white to yellow solid.


Soluble in DMSO (25mg/ml, warm) or 100% ethanol (10mg/ml, warm).

Formula: C₂₁H₂₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00210755
CAS Number: 99754-06-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ee3afaeaccc5027d7cb9976f971d008df86ecc8&vid=9d5003154d9fa9490fb85e789c48ed2d2b22b80c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR