D-erythro-Dihydrosphingosine ≥98%

Supplier: Enzo Life Sciences

Synonyms: (2S,3R)-2-Aminooctadecane-1,3-diol

BML-SL125-0010 BML-SL125-0050
89164-596EA 89.38 USD
89164-596 89164-598
D-erythro-Dihydrosphingosine ≥98%
D-erythro-Dihydrosphingosine

Sphingosine precursor


Biosynthetic precursor of sphingosine. Inhibitor of protein kinase C (PKC). Blocks phospholipases A2 (PLA2) and the D-sphingosine precursor.

Formula: C₁₈H₃₉NO₂
MW: 301.50 g/mol
MDL Number: MFCD07371371
CAS Number: 764-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d59a66d01008d6ca93b2f13390c1279a23d0e904&vid=62e3a258b169096f69670c63b89d25becbab89c1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR