N-Hexanoyl-D-erythro-sphingosine ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-SL110-0005 BML-SL110-0025
89164-576EA 91.56 USD
89164-576 89164-578
N-Hexanoyl-D-erythro-sphingosine ≥98% (by TLC)
N-Hexanoyl-D-erythro-sphingosine

A biologically active, cell-permeable, but nonphysiologic ceramide analog. It activates protein phosphatase 2A and MAP kinase (MAPK/ERK), suppresses insulin-induced tyrosine phosphorylation and inhibits glycoprotein traffic by the secretory pathway. It inhibits diacylglycerol accumulation and phospholipase D activation in fibroblasts. It induces an arrest at the G0/G1 transition of the cell cycle (10μM) and causes apoptosis (15μM or greater) in Molt-4 leukemia cells.


  • Cell-permeable ceramide analog


White waxy solid.


Soluble in 100% ethanol (>25mg/ml) and DMSO (>50mg/ml).

Formula: C₂₄H₄₇NO₃
Storage Temperature: Freezer
MDL Number: MFCD00870174
CAS Number: 124753-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR