MG-132
Supplier: Enzo Life Sciences
BML-PI102-0005 BML-PI102-0025
89161-566EA 78.48 USD
89161-566 89161-568
MG-132
Proteins and Peptides
MG-132 is a potent, cell permeable and selective proteasome inhibitor (Ki = 4nM).1 It inhibits NF-κB activation by preventing IκB degradation (IC50 = 3μM). The peptide blocks degradation of short-lived proteins, which in turn induces HSP and ER chaperone expression, leading to thermotolerance (1μM MG-132, 2 h.). It also stimulates neurite outgrowth in PC12 cells (20nM optimal). The peptide has also been reported to increase the survival rate of mesenchymal stem cells following their transplantation. IC50’s for inhibition of Suc-LLVY-AMC and Z-LLL-AMC cleaving activities of proteasome were 0.85 and 0.1μM respectively. The ubiquitin-proteasome system (UPS) and autophagy serve as two complementary, reciprocally regulated protein degradation systems, thus blockade of UPS by MG-132 activates autophagy.
  • Key inhibitor for use in proteasome research.
White solid.
Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).