Paroxetine hydrochloride ≥98%

Supplier: Enzo Life Sciences

Synonyms: (3S,4R)-3-((Benzo[d][1,3]dioxol-5-yloxy)methyl)-4-(4-fluorophenyl)piperidine hydrochloride

BML-NS710-0020 BML-NS710-0100
89160-896EA 120.99 USD
89160-896 89160-898
Paroxetine hydrochloride ≥98%
Paroxetine hydrochloride

Highly potent and selective serotonin uptake inhibitor


Paroxetine is a highly potent and selective serotonin (5-HT) uptake inhibitor. It acts by binding to the serotonin transporter (SERT) with high affinity (Ki = 05 nM). In vitro paroxetine competitively inhibits the uptake of 5-HT by rat hypothalamic and cortical synaptosomes with only very weak effects on noradrenaline and dopamine uptake. Paroxetine is in clinical use for the treatment of depression, anxiety and other disorders.

Formula: C₁₉H₂₁ClFNO₃
CAS Number: 78246-49-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba69eb5314f3125cb53b0560871815105a399410&vid=54418f322a7ec340e7218df700018900b221cdcb&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR