5-Hydroxydecanoate sodium salt ≥98% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
5-Hydroxydecanoate sodium, Sodium 5-hydroxydecanoate
BML-KC141-0100
BML-KC141-0500
89160-218EA
107.91
USD
89160-218
89160-220
5-Hydroxydecanoate sodium salt ≥98% (by TLC)
5-Hydroxydecanoate sodium salt
Inhibits ATP-sensitive K+ channels.
- K+ channel blocker
White solid.
Soluble in water (25 mg/ml).
Formula:
C₁₀H₁₉O₃·Na |
MDL Number:
MFCD00153808 CAS Number: 71186-53-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...