Pifithrin-α hydrobromide ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-GR325-0005 BML-GR325-0025
89158-928EA 106 USD
89158-928 89158-930
Pifithrin-α hydrobromide ≥98% (by TLC)
Pifithrin-alpha hydrobromide

Chemical inhibitor of p53 that protects mice from the side effects of cancer therapy. Reversibly blocks p53-dependent transcriptional activation and apoptosis. Protects against neuronal death in models of stroke and neurodegenerative disorders. Suppresses self-renewal of embryonic stem cells.


  • p53 inhibitor


White solid.


Soluble in DMSO (>25mg/ml) or methanol.

Formula: C₁₆H₁₈N₂OS·HBr
Storage Temperature: Freezer
MDL Number: MFCD00417851
CAS Number: 63208-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d270677fb1594cb6d3f46cbfb522b947cc3ab15e&vid=4237ff6b9af9bb33d19589687d711179724e9933&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR