Geldanamycin ≥98% (by TLC) (conjugated to FITC)

Supplier: Enzo Life Sciences

Synonyms: (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

BML-EI361-0001
89158-178EA 342 USD
89158-178
Geldanamycin ≥98% (by TLC) (conjugated to FITC)
Geldanamycin

A novel geldanamycin fluorescent probe that may be used in a fluorescence polarization assay for HSP90 inhibitors. May also be used for detection of cell surface HSP90.


  • HSP90 inhibitor

Formula: C₂₉H₄₀N₂O₉
MW: 560.64 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00274570
CAS Number: 30562-34-6
https://pr.vwr.com/stibo/web/std.lang.all/38/01/45403801.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR