Geldanamycin ≥98% Biotin

Supplier: Enzo Life Sciences

Synonyms: (4E,6Z,8S,9S,10E,12S,13R,14S,16R)-13-Hydroxy-8,14,19-trimethoxy-4,10,12,16-tetramethyl-3,20,22-trioxo-2-azabicyclo[16.3.1]docosa-1(21),4,6,10,18-pentaen-9-yl carbamate

BML-EI341-0001
89158-122EA 297 USD
89158-122
Geldanamycin ≥98% Biotin
Geldanamycin

A useful tool for affinity purification of HSP-90 and HSP-90 dependent client proteins.


  • HSP90 inhibitor

Formula: C₂₉H₄₀N₂O₉
MW: 560.64 g/mol
MDL Number: MFCD00274570
CAS Number: 30562-34-6
https://pr.vwr.com/stibo/web/std.lang.all/37/86/45403786.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR