(±)-Blebbistatin 98

Supplier: Enzo Life Sciences

Synonyms: 3a-Hydroxy-6-methyl-1-phenyl-1,2,3,3a-tetrahydro-4H-pyrrolo[2,3-b]quinolin-4-one, Blebbistatin

BML-EI315-0005 BML-EI315-0025
89158-050EA 179.85 USD
89158-050 89158-052
(±)-Blebbistatin 98
(±)-Blebbistatin

Inhibitor of non-muscle myosin II


Blebbistatin (so named because of its ability to block cell blebbing) is a selective and high-affinity (IC50 approx. 4 µM) inhibitor of non-muscle myosin II. During cell division blebbistatin inhibits contraction of the cleavage furrow without disrupting mitosis. It inhibits pancreatic adenocarcinoma cellular invasiveness and PDGF-stimulated fibroblast-collagen matrix contraction.

Formula: C₁₈H₁₆N₂O₂
MW: 292.33 g/mol
MDL Number: MFCD04113775
CAS Number: 674289-55-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31ca389dbe4898ab893222576a6838f8ad8435d4&vid=d5829c16d9bbb59770a74b85086a93187cc5ae09&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR