CL-82198 (MMP-13 inhibitor) ≥98% (by TLC)

Supplier: Enzo Life Sciences

Synonyms: N-[4-(4-morpholinyl)butyl]-2-benzofurancarboxamide, CL-82198, N-(4-Morpholinobutyl)benzofuran-2-carboxamide

BML-EI302-0005 BML-EI302-0025
89158-006EA 97.01 USD
89158-006 89158-008
CL-82198 (MMP-13 inhibitor) ≥98% (by TLC)
CL-82198 (MMP-13 inhibitor)

A novel and selective inhibitor of MMP-13 (89% at 33 µM). Acts via binding to the S1' pocket of MMP-13 without metal chelation.


  • MMP inhibitor


White solid.


Soluble in water (25mg/ml).

Formula: C₁₇H₂₂N₂O₃
MW: 302.37 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12828877
CAS Number: 307002-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b64309f5643347c61a5c042fd751405b920fdb74&vid=72e825ea04d3685a86e3cf07f7a108540eae3fd0&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR