ZM-336372 ≥95% (by HPLC)
Supplier: Enzo Life Sciences
Synonyms:
3-(Dimethylamino)-N-(3-(4-hydroxybenzamido)-4-methylphenyl)benzamide, ZM 336372
BML-EI298-0005
BML-EI298-0025
89157-998EA
207.1
USD
89157-998
89158-000
ZM-336372 ≥95% (by HPLC)
ZM-336372
Potent and specific inhibitor of the protein kinase c-Raf (IC50=70nM). Has no significant effect on many other protein kinases tested (even at 50µM) with the exception of p38 (SAPK2a) (IC50=2µM) and SAPK2b (p38β) (IC50=2µM). Induces a paradoxical >100-fold activation of c-Raf in whole cells.
- Raf inhibitor
White to off-white solid.
Soluble in DMSO (200mg/ml).
Formula:
C₂₃H₂₃N₃O₃ MW: 389.455 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD02683971 CAS Number: 208260-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...