Tranylcypromine ≥99%

Supplier: Enzo Life Sciences

BML-EI217-0001 BML-EI217-0005
89157-800EA 163.5 USD
89157-800 89157-802
Tranylcypromine ≥99%
Tranylcypromine

Demethylase inhibitor


Inhibits prostacyclin synthase. Monoamine oxidase inhibitor. Effective small molecule inhibitor of histone demethylation. Inhibits the demethylase BHC110/LSD1 (IC50<2μM).

Formula: C₉H₁₁N
MW: 133.2 g/mol
MDL Number: MFCD00001302
CAS Number: 155-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c16e66bda895b8546fd5f1ad623badb6050b02f&vid=e127bc1a06fb16d129509ae6154afdb1b83bd6fe&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR