1-Oleoyl-2-acetyl-sn-glycerol ≥98% (by TLC)

Supplier: Enzo Life Sciences

BML-DG125-0010 BML-DG125-0050
89157-354EA 171.13 USD
89157-354 89157-356
1-Oleoyl-2-acetyl-sn-glycerol ≥98% (by TLC)
1-Oleoyl-2-acetyl-sn-glycerol

PKC activator.


  • Formulation: Colorless oil or waxy solid.


In vitro potent activator of protein kinase C (PKC). Stimulator of secretion and aggregation in blood platelets. Stimulates formation of actin nucleation sites. Stimulation of pp60c-src phosphorylation in EBV transformed B-cells, 470 μM.


Soluble in DMSO or ethanol.

Formula: C₂₃H₄₂O₅
MW: 398.6 g/mol
Storage Temperature: Freezer
CAS Number: 86390-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27cef47d2ba3487cf240bfb7cf937362b0af86b7&vid=401a937f48e726b3e79b4029bd49e320543c97f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Source Synthetic
Purity ≥98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR