8-(4-Chlorophenylthio)adenosine-3',5'-cyclic monophosphate sodium salt (synthetic) ≥96% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 8-CPT cAMP

BML-CN130-0020 BML-CN130-0100
89157-010EA 94.83 USD
89157-010 89157-012
8-(4-Chlorophenylthio)adenosine-3',5'-cyclic monophosphate sodium salt (synthetic) ≥96% (by HPLC)
8-(4-Chlorophenylthio)adenosine-3',5'-cyclic monophosphate sodium salt

PKA activator.


  • Formulation: White solid.


A lipophilic, cell-permeable cAMP analog which activates cAMP- and cGMP-dependent protein kinase. More potent than cAMP and more resistant to phosphodiesterasese, with good selectivity for site B of PKA. For specific activation of the cAMP pathway the highly membrane permeable Sp-5,6-DCI-cBIMPS.


Soluble in water (25 mg/mL).

Formula: C₁₆H₁₄ClN₅NaO₆PS
Storage Temperature: Freezer
MDL Number: MFCD00057683
CAS Number: 93882-12-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61a15d256ed42f00776dfbda41e6b9fb3798e4c0&vid=8732bd1faee52be47103594a86ff97fc3159856c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Source Synthetic
Purity ≥96%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR