Dibutyryl-cAMP sodium salt ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Bucladesine sodium salt, DC-2797, Dibutyl cyclic AMP sodium salt, Dibutyryl-cAMP, N6,2'-O-Dibutyryladenosine 3',5'-Cyclic monophosphate sodium salt, DbcAMP, Sodium (4aR,6R,7R,7aR)-6-(6-butyramido-9H-purin-9-yl)-7-(butyryloxy)tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-2-olate 2-oxide, Bucladesine

BML-CN125-0100 BML-CN125-0030
89157-008EA 179.85 USD
89157-008 76002-060
Dibutyryl-cAMP sodium salt ≥98% (by HPLC)
Dibutyryl-cAMP sodium salt

PKA activator.


  • Formulation: White crystalline solid.


A cell-permeable cAMP analog which preferentially activates cAMP-dependent protein kinase. The products releaes butyrate due to intracellular and extracellular esterase action. Butyrate was shown to have distinct biological effects.


Soluble in water (100 mg/mL), DMSO (200 mM) or 96% ethanol (25 mM).


Synonym: Dibutyl cyclic AMP sodium salt.

Formula: C₁₈H₂₃N₅O₈P·Na
MW: 491.37 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00005843
CAS Number: 16980-89-5
EINECS: 241-059-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01751242474e209b481659378043656b9f7cae4e&vid=174f765272dc5542a1079f769a8709dfa0a865b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (HPLC)
Identity Determined by UV spectrum

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR