Mdivi -1 ≥97% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: Mdivi-1, 3-(2,4-Dichloro-5-methoxyphenyl)-2-mercaptoquinazolin-4(3H)-one, 3-(2,4-Dichloro-5-methoxyphenyl)-2-thioxo-2,3-dihydro-4(1H)-quinazolinone

BML-CM127-0010 BML-CM127-0050
89156-980EA 126.44 USD
89156-980 89156-982
Mdivi -1 ≥97% (by HPLC)
Mdivi -1

Mdivi-1 is a selective inhibitor of mitochondrial division in yeast and mammalian cells which acts via inhibiting the mitochondrial division dynamin. In cells, Mdivi-1 inhibits apoptosis by inhibiting mitochondrial outer membrane permeabilization. Mdivi-1 causes the rapid (50= ~10µM. In yeast, time-lapse fluorescence microscopy revealed no detectable mitochondrial division after treatment with Mdivi-1. It inhibits Dnm1 GTPase activity in a dose dependent manner with an estimated IC50 of 1-10µM but is not a general inhibitor of GTPases.


  • Inhibitor of mitochondrial division


White solid.


Soluble in DMSO (25mg/ml).

Formula: C₁₅H₁₀Cl₂N₂O₂S
MW: 353.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00974506
CAS Number: 338967-87-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4688b457560944b7e382e87d17432f4fc20c55f0&vid=95aa8482089ea2c49a6d34947af4638c96dc8924&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR