Oligomycin A ≥97% (by HPLC, TLC)

Supplier: Enzo Life Sciences

Synonyms: (1R,2'R,4E,5'S,6S,6'S,7R,8S,10R,11R,12S,14R,15S,16R,18E,20E,22R,25S,28S,29R)-22-Ethyl-7,11,14,15-tetrahydroxy-6'-((R)-2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-3',4',5',6'-tetrahydro-2,26-dioxaspiro[bicyclo[23.3.1]nonacosa[4,18,20]triene-27,2'-pyran]-3,9,13-trione

BML-CM111-0005 BML-CM111-0025
89156-934EA 244.16 USD
89156-934 89156-936
Oligomycin A ≥97% (by HPLC, TLC)
Oligomycin A

Inhibits mitochondrial ATPases (F1F0)


  • White solid
  • Soluble in DMSO (300 mg/mL), 100% ethanol (200 mg/mL) or acetone (800 mg/mL)


Stimulates lysosome acidification due to shunting of membrane potential. Induces apoptosis in a variety of cell types. Induces autophagy in the IPLB-LdFB insect cell line.

Formula: C₄₅H₇₄O₁₁
Storage Temperature: Freezer
MDL Number: MFCD00065705
CAS Number: 579-13-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ffbbed499f16bb904741f475a6682dc0ec0d027&vid=961023b03ced8efe12a3d92f01d30cfcdb561874&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR