5-Nitro-2-(3-phenylpropylamino)benzoic acid ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 5-Nitro-2-((3-phenylpropyl)amino)benzoic acid, NPPB

BML-CM100-0010 BML-CM100-0050
89156-924EA 70.85 USD
89156-924 89156-926
5-Nitro-2-(3-phenylpropylamino)benzoic acid ≥98% (by HPLC)
5-Nitro-2-(3-phenylpropylamino)benzoic acid

A potent chloride channel blocker (IC50=0.1-100 µM depending on channel subtype and assay method). Inhibits cyclooxygenase (COX; IC50=8 µM). Evidence of mitochondrial uncoupling in mouse peritoneal macrophages, due to protonophore activity.


  • Cl- channel blocker


Yellow solid.


Soluble in DMSO (25mg/ml) or 100% ethanol (10mg/ml; warm).

Formula: C₁₆H₁₆N₂O₄
MW: 300.314 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00153851
CAS Number: 107254-86-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7f5d22ef38eef0ef5e994d5985fcd6e2cdba19f&vid=c1af03ff226086140cd085d181c009de0fae85de&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR