(R)-Roscovitine ≥95% (by TLC)
Supplier: Enzo Life Sciences
Synonyms:
2-(R)-(1-Ethyl-2-hydroxyethylamino)-6-benzylamino-9-isopropylpurine, Roscovitine, (R)-2-((6-(Benzylamino)-9-isopropyl-9H-purin-2-yl)amino)butan-1-ol
BML-CC205-0005
BML-CC205-0002
89156-904EA
246.34
USD
89156-904
76002-058
(R)-Roscovitine ≥95% (by TLC)
(R)-Roscovitine
CDK inhibitor
Potent and selective inhibitor of CDKs. Selective for CDK1/cyclin B kinase (IC50=450nM) [2], CDK2 (IC50=700nM) [4] and CDK5/p35 (IC50=160nM) [4,5]. More potent than olomoucine (Prod. No. BML-CC200). Inhibits M phase promoting factor (MPF) kinase activity. Has effects on calcium channel gating.
Formula:
C₁₉H₂₆N₆O MW: 354.45 g/mol |
MDL Number:
MFCD02266401 CAS Number: 186692-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...