AMG-9810 ≥98%

Supplier: Enzo Life Sciences
Warning

Synonyms: (E)-3-(4-(tert-butyl)phenyl)-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylamide, AMG9810, (E)-3-(4-(tert-Butyl)phenyl)-N-(2,3-dihydrobenzo[b][1,4]dioxin-6-yl)acrylamide, AMG 9810, (2E)-N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-[4-(1,1-dimethylethyl)phenyl]-2-propenamide, 2E-N-(2,3-Dihydro-1,4-benzodioxin-6-yl)-3-[4-(1,1-dimethylethyl)phenyl]-2-Propenamide

BML-CA232-0010 BML-CA232-0050
89156-800EA 124.26 USD
89156-800 89156-802
AMG-9810 ≥98%
AMG-9810

TRPV1 antagonist.


  • Formulation: Off-white solid.


AMG-9810 is an antagonist of capsaicin activation of TRPV1 (IC₅₀ = 24.5 nM) as well as activation by acid, heat, and endogenous ligands.


Soluble in DMSO (>25 mg/mL), ethanol (20 mg/mL).

Formula: C₂₁H₂₃NO₃
MW: 337.41 g/mol
MDL Number: MFCD00955103
CAS Number: 545395-94-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69598676fb4238cd5423643b66ba6d6ee1204447&vid=329b053ef884e112bc4594cb0598b626c88fcbe9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR