Amlodipine ≥98%

Supplier: Enzo Life Sciences

Synonyms: 3-Ethyl 5-methyl 2-((2-aminoethoxy)methyl)-4-(2-chlorophenyl)-6-methyl-1,4-dihydropyridine-3,5-dicarboxylate

BML-CA202-0100 BML-CA202-1000
89156-768EA 97.01 USD
89156-768 89156-770
Amlodipine ≥98%
Amlodipine

Ca²⁺ channel blocker.


  • Formulation: Almost white crystal powder


Amlodipine is a third generation L-type calcium channel blocker with a unique spectrum of actions, over and above its antihypertensive activity which include stimulation of NO synthase, inhibition of monocyte-endothelial adhesion, inhibition of oxidative stress and sustained duration of action.


Soluble in methanol, DMSO (25 mg/mL), 100% ethanol (25 mg/mL), dimethyl formamide (25 mg/mL) or chloroform.

Formula: C₂₀H₂₅ClN₂O₅
MW: 408.879 g/mol
MDL Number: MFCD00864687
CAS Number: 88150-42-9
EINECS: 425-820-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=303176c0b9280c8ebda9a64fd6ad0b6faad16a58&vid=4384faee3683a8aa4d67f515d6bbeb8b3e57b3dd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥98% (Assay)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR