1,5-Isoquinolinediol 98
Supplier: Enzo Life Sciences
Synonyms:
5-Hydroxyisoquinoline-1(2H)-one
BML-AP102-0020
BML-AP102-0100
89156-568EA
83.93
USD
89156-568
89156-570
1,5-Isoquinolinediol 98
1,5-Isoquinolinediol
PARP inhibitor
Potent inhibitor of inducible nitric oxide synthase (iNOS; NOS II) in mouse macrophages. Potent and selective PARP inhibitor. Inhibits nitric oxide (SIN-1) induced PARP activation in rat hepatocytes (at 20µM). Neuroprotectant.
Formula:
C₉H₇NO₂ MW: 161.16 g/mol |
MDL Number:
MFCD00006900 CAS Number: 5154-02-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...