Ryania 99 HPLC_ASSAY_METHOD, high_purity

Supplier: Enzo Life Sciences
Warning

Synonyms: Ryanodine

630-062-M001 630-062-M005
89152-958EA 226.72 USD
89152-958 89152-960
Ryania 99 HPLC_ASSAY_METHOD, high_purity
Ryania

Ca²⁺ channel activator.


  • Formulation: White to off-white solid.


Induces the release of intracellular Ca²⁺. Ryanodine receptors mediate adenine nucleotide, Ca²⁺ and caffeine-sensitive release of Ca²⁺ from sarcoplasmic reticulum of cardiac and skeletal muscle and may also mediate intracellular Ca²⁺ release in neurons. Biphasic effect on sarcoplasmic reticulum calcium channels, nanomolar levels open the channel and micromolar levels close it.


Soluble in methanol or DMSO (>29 mg/mL), in 100% ethanol (16 mg/mL), slightly soluble in water (warm).

Formula: C₂₅H₃₅NO₉
MW: 493.553 g/mol
Storage Temperature: Freezer
CAS Number: 15662-33-6
EINECS: 239-732-2
UN: 3077
ADR: 9 III
Merck Index: 14,08307
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87362d4cf4716a6b82e6b5679afd495448634939&vid=a04bc154773276a9ae13f757819648f14d809862&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Source Isolated from Ryania speciosa, Flacourtiaceae
Purity ≥99% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR