MPLA (from Salmonella minnesota) ≥99.9%, TLRgrade™

Supplier: Enzo Life Sciences

Synonyms: Monophosphoryl Lipid A

581-202-L001 581-202-R500
89152-794EA 604.95 USD
89152-794 76002-220
MPLA (from Salmonella minnesota) ≥99.9%, TLRgrade™
MPLA

TLR4 activator.


Isolated from Salmonella minnesota R595 (Re mutant). Prepared by acid hydrolysis from purified Salmonella minnesota R595 (Re) LPS (isolated by a modification of the PCP extraction method), converted to the uniform salt form.


Activates Toll-like receptor (TLR) 4. Does not activate TLR2 as determined with splenocytes and macrophages from TLR4 deficient mice. Exhibits reduced endotoxic and pyrogenic activity compared to lipid A and LPS.


Synonym: MPL-A from Salmonella minnesota R595 (Re).

Order Now

Specification Test Results

Concentration 1 mg/ml
Formulation Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water
Purity ≥99.9% (regarding protein or DNA contaminants, KDO not detectable)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR