Lipopolysaccharides (from S. abortus equi) S-form, TLRgrade™ ready-to-use
Supplier: Enzo Life Sciences
|
Danger
|
TLR4 activator.
Smooth (S)-form LPS, isolated and purified from Salmonella abortus equi by modification of the phenol water extraction and PCP method, converted to the uniform salt form.
Strong activator of Toll-like receptor (TLR) 4. Does not activate TLR2 or other TLRs as determined with splenocytes and macrophages from TLR4 deficient mice. No further re-extraction required. Smooth (S)-form LPS are commonly the preferred choice for whole animal studies, whereas Rough (R)-form LPS are primarily used in cellular in vitro activation studies.
Specification Test Results
Concentration | 1 mg/ml |
Formulation | Liquid. Sterile, ready-to-use solution in pyrogen-free double distilled water |
Purity | Absence of detectable protein or DNA contaminants with agonistic TLR activity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...