MTEP ≥98% (by NMR)

Supplier: Enzo Life Sciences

550-381-M005 550-381-M025
89152-646EA 126.44 USD
89152-646 89152-648
MTEP ≥98% (by NMR)
MTEP

mGluR5 antagonist.


  • Formulation: White to yellow solid.


Potent and highly selective antagonist for mGluR5 with a 5-fold higher anxiolytic activity than MPEP. Shows fewer off-target effects than MPEP.


Soluble in DMSO, 100% ethanol or methanol.

Formula: C₁₁H₈N₂S
Storage Temperature: Refrigerator
CAS Number: 329205-68-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d8274421d5f6fa2e876fe131e9c90290a1e8939&vid=80482b917d04891d80af49130a17ad530b08608d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (NMR) ≥98%
Identity Identity determined by ¹H-NMR
Appearance Pale yellow oil

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR