Flavopiridol ≥97% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 2-(2-Chlorophenyl)-5,7-dihydroxy-8-((3S,4R)-3-hydroxy-1-methylpiperidin-4-yl)-4H-chromen-4-one

430-161-M005 430-161-M025
89151-284EA 105.73 USD
89151-284 89151-286
Flavopiridol ≥97% (by HPLC)
Flavopiridol

CDK inhibitor


Potent inhibitor of cyclin-dependent kinases (CDKs). Induces apoptosis in certain tumor cells. Interacts with multidrug resistance protein 1 (MRP1).


Synthetic.

Formula: C₂₁H₂₀ClNO₅
MW: 401.840 g/mol
MDL Number: MFCD20501884
CAS Number: 146426-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d810bf441d6df51089a10316043e34eed91ff04&vid=5b4ac6cba26c9e6620b75adba61115099981d8b3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR