NOR-3 ≥97% (by HPLC)
Supplier: Enzo Life Sciences
430-011-M005 430-011-M010
89150-862EA 101.37 USD
89150-862 89150-864
NOR-3 ≥97% (by HPLC)
NOR-3
Nitric oxide donor
Non-thiol-based nitric oxide (NO) donor. NOR-3 shows a variety of biological and pharmacological effects. Strong vasodilatory effects in rat aorta, rabbit aorta and dog coronary artery; potent inhibitory activities against antiplatelet aggregation and thrombus formation; effective aggregation inhibition of human platelet raising by ADP; cardioprotective effects in the ischemia/reperfusion system; antianginal effect.
CAS Number: 163180-49-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c8cbdc9262ee3de34a9b77697371a0c66984d5e&vid=7159d2ffbc3679395cdedd4615952ad05d10336d&prid=web&clid=SAPDAM