DMNQ ≥99% (by TLC)

Supplier: Enzo Life Sciences
Warning

Synonyms: 2,3-Dimethoxy-1,4-naphthoquinone, 2,3-Dimethoxynaphthalene-1,4-dione

420-027-M005 420-027-M010 420-027-M025
89150-740EA 124.26 USD
89150-740 89150-742 89150-744
DMNQ ≥99% (by TLC)
DMNQ

Inducer of intracellular ROS generation.


  • Formulation: Yellow crystalline solid.


Redox-cycling agent that induces intracellular superoxide anion formation and, depending on the concentration, induces cell proliferation, apoptosis or necrosis. DMNQ does not react with free thiol groups, is non-alkylating and adduct-forming in contrast to other quinones. Thus, DMNQ is a valuable tool for the generation of reactive oxygen species (ROS) in order to study the role of ROS in cell toxicity, apoptosis and necrosis.


Soluble in acetone, DMSO or methanol.


Synonym: 2,3-Dimethoxy-1,4-naphthoquinone

Formula: C₁₂H₁₀O₄
MW: 218.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00184331
CAS Number: 6956-96-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c23601224d88e9c723f6b1f7916550362ca43e7b&vid=464b144407bb3bef8a1bcf2562b22a3d17cff0ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity ≥99% (TLC)
Identity Determined by NMR

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR