Amidepsine A ≥95% (by HPLC)

Supplier: Enzo Life Sciences

380-262-M001 380-262-MM25
89150-286EA 407.66 USD
89150-286 89150-288
Amidepsine A ≥95% (by HPLC)
Amidepsine A

DGAT inhibitor


Inhibitor of diacylglycerol acyltransferase (DGAT). Shows inhibitory effect in rat liver microsomes (IC50=10.2µM) and Raji cells (IC50=15.5µM). Excessive accumulation of triacetylglycerol produced by DGAT may cause fatty liver, obesity and hypertriglyceridemia, which may lead to atherosclerosis, diabetes and metabolic disorders.


Isolated from fungal strain FO-2943.


Información para pedidos: Fungal strain courtesy of the Kitasato Institute, Tokyo.

Formula: C₂₉H₂₉NO₁₁
CAS Number: 169181-28-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f728e0ac9b6226e8939fe33d3406be16c88526f6&vid=a12c65965645250d74c47d3abde109cc2e3c0fe3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR