Gentamicin sulphate (from Micromonospora sp.)

Supplier: Enzo Life Sciences

Synonyms: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

380-003-G001 380-003-G005 380-003-G010 380-003-G025
89149-782EA 73.03 USD
89149-782 89149-784 89149-786 89149-788
Gentamicin sulphate (from Micromonospora sp.)
Gentamicin sulphate

White to off-white powder


Broad-spectrum cell culture antibiotic that is nontoxic to viruses and mammalian cells at antibacterial and antimycoplasmal concentrations. Due to its extended stability and slow development of bacterial resistance, it is a useful antibiotic in long-term virus und tissue culture studies.


Soluble in water, almost insoluble in alcohol or ether


Envase: Shipping temperature: Ambient

Storage Temperature: Refrigerator
MDL Number: MFCD00270181
CAS Number: 1405-41-0
Merck Index: 13,04403
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a61c37f60c6050e40327cf9242920a778786522&vid=66dffcae0bee3ed1590fce327a079cbfeed733ee&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR