Microcystis aeruginosa Microcystin LY
Supplier: Enzo Life Sciences
350-148-C025
350-148-C100
350-148-M001
89149-172EA
244.16
USD
89149-172
89149-174
76417-958
Microcystis aeruginosa Microcystin LY
Proteins and Peptides
Analog of microcystin-LR (Prod. No. ALX-350-012) with Tyr substituted in place of Arg. Its toxicity is intermediate between the more toxic microcystin-LF and the less toxic microcystin-LR.
Isolated from Microcystis aeruginosa.