Nodularin ≥95% (by HPLC)

Supplier: Enzo Life Sciences

350-061-C100 350-061-C250 350-061-M001
89148-992EA 171.13 USD
89148-992 89148-994 89148-996
Nodularin ≥95% (by HPLC)
Nodularin

Inhibitor of PP1 and PP2A


Inhibitor of protein phosphatase 1 (PP1) (IC50=1.8nM), protein phosphatase 2A (PP2A) (IC50=0.026nM) and to a lesser extent protein phosphatase 2B (PP2B) (IC50=8.7µm). Similar to microcystin-LR (Prod. No. ALX-350-012) but with increased water solubility. Genotoxic. Hepatoxic. Induces oxidative DNA damage by oxidation of purines. Induces apoptosis in HepG2 cells.


Isolated from Nodularia spumigena.

Formula: C₄₁H₆₀N₈O₁₀
MW: 824.977 g/mol
CAS Number: 118399-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6203e66a9c48e0e921fe8a489237689ef78e33a2&vid=0957e70c1c4532db120d794200ef4d3020e86ced&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR