Hypericin (native) ≥98% (by HPLC)

Supplier: Enzo Life Sciences

Synonyms: 1,3,4,6,8,13-Hexahydroxy-10,11-dimethylphenanthro[1,10,9,8-opqra]perylene-7,14-dione P-conformer

350-030-M001 350-030-M005 350-030-M010
89148-950EA 97.01 USD
89148-950 89148-952 89148-954
Hypericin (native) ≥98% (by HPLC)
Hypericin

PKC inhibitor


A potent and selective inhibitor of protein kinase C (IC50=3.4µM). Anti-viral and anti-retroviral actitity. Displays antineoplastic and antitumor actitities. Most activities require light activation. Induces apoptosis in neuroblastoma cells at 1µM or above (IC50=0.1µM at 29 mW/cm2 (tungsten light)). Inhibitition of other kinases (e.g. EGFR-PTK, CKII and MAP kinase). Bright red fluorescence emissino and photostability. Antidepressant.


Isolated from Hypericum perforatum.

Formula: C₃₀H₁₆O₈
MW: 504.45 g/mol
MDL Number: MFCD00016683
CAS Number: 548-04-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f0c2aa5dde97a5430a95d7ddd9221fb04712e800&vid=009125525d0f645e4b60691bad17e1bbaca31fcd&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR