Curcumin (from Curcuma longa) ≥98% (by HPLC), high purity

Supplier: Enzo Life Sciences

Synonyms: (1E,6E)-1,7-Bis(4-hydroxy-3-methoxyphenyl)hepta-1,6-diene-3,5-dione

350-028-M010 350-028-M050 350-028-M250
89148-944EA 50.14 USD
89148-944 89148-946 89148-948
Curcumin (from Curcuma longa) ≥98% (by HPLC), high purity
Curcumin

Inhibitor of Lipoxygenase and COX


Dual inhibitor of 5-lipoxygenase (IC50=8µM) and cyclooxygenase (COX) (IC50=52µM). Also inhibits glutathione S-transferase, induction of nitric oxide (NO) in activated macrophages and inhibits EGF-induced tyrosine phosphorylation of EGF receptors. Antioxidant. Inhibitor of NAD(P)H:quinone oxidoreductase 1 (NQO1), disrupting the binding of NQO1 to wild type p53 inducing ubiquitin-independent degradation of p53 and inhibits p53-mediated apoptosis in normal thymocytes and myeloid leukemic cells. Has been shown to inhibit NF-κB, possibly through inhibition of JNK (IC50=10µM), the COP9 signalosome kinase (IC50=10µM) and p300/CBP.

Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
MDL Number: MFCD00008365
CAS Number: 458-37-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f84ca05ac290ddf79b20e009b972e6c6274a4d45&vid=513a48203fd0ec0d5dcdd033cbe9c4e0ab7013b0&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR