L-Canavanine sulfate 99 TLC_ASSAY_METHOD

Supplier: Enzo Life Sciences

Synonyms: Canavanine sulphate, L-α-Amino-γ-(guanidinooxy)-n-butyric acid sulphate

350-002-M100 350-002-M500
89148-884EA 57.77 USD
89148-884 89148-886
L-Canavanine sulfate 99 TLC_ASSAY_METHOD
L-Canavanine sulfate hydrate

The non-protein amino acid L-canavanine is an analog of L-arginine. Selective inducible nitric oxide synthase (iNOS; NOS II) inhibitor. Induces apoptotic cell death and shows antiproliferative and immunotoxic effects.


  • iNOS inhibitor


White to-off white powder.


Soluble in water (100mg/ml).

Formula: C₅H₁₂N₄O₃·H₂SO₄
MW: 274.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00012618
CAS Number: 2219-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd013f9120648eb73c5163f977d0a13dfd5c630e&vid=14ad7e3f001754e6ec63920147c75b476f05cd05&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR