Imatinib mesylate

Supplier: Enzo Life Sciences

270-492-M500 270-492-M025
89148-084EA 559 USD
89148-084 76203-094
Imatinib mesylate
Imatinib mesylate

Tyrosine kinase inhibitor


Tyrosine kinase inhibitor. Highly specific for BCR-ABL, the enzyme associated with chronic myelogenous leukemia (CML) and certain forms of acute lymphoblastic leukemia (ALL). Also shown to inhibit the transmembrane receptor kit and platelet-derived growth factor (PDGF) receptors.

Formula: C₂₉H₃₁N₇O·CH₄SO₃
MDL Number: MFCD04307699
CAS Number: 220127-57-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c279d3d3260c5978d1e536b10cee8d573d36e37&vid=c4f8d33c9e3c0cae97c7b7195ea932baa6de05aa&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR