NU6102 ≥95% (by HPLC)

Supplier: Enzo Life Sciences

270-419-M001 270-419-M005
89147-852EA 106.82 USD
89147-852 89147-854
NU6102 ≥95% (by HPLC)
NU6102

CDK/cyclin inhibitor


1,000-fold more potent inhibitor of CDK1/cyclin B (IC50=9.5nM) and CDK2/cyclin A3 (IC50=5.4nM) than NU2058 (Prod. No. ALX-270-394). Selective for CDK1 and CDK2 compared to CDK4/D1 (IC50=1.6µM), DYRK1A (IC50=0.9µM), PDK1 (IC50=0.8µM) and ROCKII (IC50=0.6µM).

Formula: C₁₈H₂₂N₆O₃S
CAS Number: 444722-95-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1acbc287c109fa9e5529a215aaa95c0e99af3ecc&vid=f28155c942011b20c3dca80761fb4512b0ab2e10&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR