SH-5
Supplier: Enzo Life Sciences
270-349-M001
270-349-MC05
89147-670EA
389.13
USD
89147-670
89147-672
SH-5
SH-5
Akt inhibitor
Inhibitor of Akt (PKB) activation without affecting activation of the upstream kinase PDK-1, or other kinases downstream of Ras such as MAPK. Induces apoptosis and kills a variety of cancer cell lines containing high levels of active Akt and depending upon Akt for survival.
Formula:
C₂₉H₅₉O₁₀P MW: 598.75 g/mol |
MDL Number:
MFCD08703608 CAS Number: 701976-54-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...